PRT-060318 98%

Supplier: Ambeed
Warning

Synonyms: 2-(((1R,2S)-2-Aminocyclohexyl)amino)-4-(m-tolylamino)pyrimidine-5-carboxamide

A424910-10MG A424910-1MG A424910-5MG
77102-178EA 48.81 USD
77102-178 77102-180 77102-182
PRT-060318 98%
PRT-060318

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₆O
MW: 340.4 g/mol
MDL Number: MFCD28155088
CAS Number: 1194961-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR