(S)-4-Benzyl-5,5-dimethyloxazolidin-2-one 98%

Supplier: Ambeed
Warning

A423369-100MG A423369-1G A423369-250MG A423369-5G
77102-160EA 44 USD
77102-160 77102-162 77102-164 77102-166
(S)-4-Benzyl-5,5-dimethyloxazolidin-2-one 98%
(S)-4-Benzyl-5,5-dimethyloxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD00674020
CAS Number: 168297-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR