4-Methoxycarbonyl-2,3-dihydro-1H-indole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl indoline-4-carboxylate
A422292-250MG
A422292-5G
A422292-1G
77102-148EA
85.94
USD
77102-148
77102-150
77686-160
4-Methoxycarbonyl-2,3-dihydro-1H-indole 97%
4-Methoxycarbonyl-2,3-dihydro-1H-indole
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₂ MW: 177.2 g/mol |
MDL Number:
MFCD07371619 CAS Number: 155135-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...