tert-Butyl 4-(4-chlorophenyl)-4-cyanopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A420904-1G A420904-250MG A420904-5G
77102-132EA 193.09 USD
77102-132 77102-134 77613-294
tert-Butyl 4-(4-chlorophenyl)-4-cyanopiperidine-1-carboxylate 97%
tert-Butyl 4-(4-chlorophenyl)-4-cyanopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁ClN₂O₂
MW: 320.81 g/mol
MDL Number: MFCD11227117
CAS Number: 218451-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR