1-(3-Chlorophenyl)-3-methyl-1H-pyrazole-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A420737-100MG
A420737-1G
A420737-250MG
77102-124EA
60.48
USD
77102-124
77102-126
77102-128
1-(3-Chlorophenyl)-3-methyl-1H-pyrazole-5-carboxylic acid 97%
1-(3-Chlorophenyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₉ClN₂O₂ MW: 236.65 g/mol |
MDL Number:
MFCD08446267 CAS Number: 1020240-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...