GDC-0623 99+%

Supplier: Ambeed
Warning

A420396-10MG A420396-1MG A420396-2MG A420396-50MG A420396-5MG
77102-110EA 115.64 USD
77102-110 77102-112 77102-114 77102-116 77102-118
GDC-0623 99+%
GDC-0623

Precaución: for Research Use Only

Formula: C₁₆H₁₄FIN₄O₃
MW: 456.21 g/mol
MDL Number: MFCD25976760
CAS Number: 1168091-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR