(9H-Fluoren-9-yl)methyl (3-aminopropyl)carbamate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-1,3-propanediamine hydrochloride

A418445-250MG A418445-25G A418445-5G A418445-1G A418445-100G
77102-106EA 6 USD
77102-106 77572-304 77572-306 77613-284 77901-299
(9H-Fluoren-9-yl)methyl (3-aminopropyl)carbamate hydrochloride 95%
(9H-Fluoren-9-yl)methyl (3-aminopropyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₁ClN₂O₂
MW: 332.82 g/mol
MDL Number: MFCD00830742
CAS Number: 210767-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR