4-Chloro-2-fluorobenzohydrazide 95%

Supplier: Ambeed
Warning

A417696-10G A417696-250MG
77102-102EA 529 USD
77102-102 77102-104
4-Chloro-2-fluorobenzohydrazide 95%
4-Chloro-2-fluorobenzohydrazide

Precaución: for Research Use Only

Formula: C₇H₆ClFN₂O
MDL Number: MFCD09813752
CAS Number: 1016768-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR