1-Amino-2-naphthonitrile 98%

Supplier: Ambeed
Warning

A417494-100MG A417494-1G A417494-250MG
77102-096EA 16 USD
77102-096 77102-098 77102-100
1-Amino-2-naphthonitrile 98%
1-Amino-2-naphthonitrile

Precaución: for Research Use Only

Formula: C₁₁H₈N₂
MDL Number: MFCD09965958
CAS Number: 3100-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR