Phenyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 95+%

Supplier: Ambeed
Warning

A415930-1G A415930-250MG A415930-5G
77102-078EA 127 USD
77102-078 77102-080 77102-082
Phenyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 95+%
Phenyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₂₂BNO₄
MDL Number: MFCD20527213
CAS Number: 756520-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR