6-(Difluoromethyl)-3-pyridinecarboxylic acid 95%

Supplier: Ambeed
Warning

A414518-100MG A414518-1G A414518-250MG
77102-066EA 81.69 USD
77102-066 77102-068 77102-070
6-(Difluoromethyl)-3-pyridinecarboxylic acid 95%
6-(Difluoromethyl)-3-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.11 g/mol
MDL Number: MFCD16621992
CAS Number: 913091-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR