o-Tolyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

A413150-25G A413150-5G
77102-044EA 96 USD
77102-044 77102-046
o-Tolyl trifluoromethanesulfonate 95%
o-Tolyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₈H₇F₃O₃S
MDL Number: MFCD29089375
CAS Number: 66107-34-4
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR