Methyl 4,5-diamino-5-oxopentanoate hydrochloride 95%

Supplier: Ambeed
Warning

A409909-100MG A409909-250MG
77102-014EA 96.55 USD
77102-014 77102-016
Methyl 4,5-diamino-5-oxopentanoate hydrochloride 95%
Methyl 4,5-diamino-5-oxopentanoate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃ClN₂O₃
MW: 196.63 g/mol
MDL Number: MFCD29917025
CAS Number: 1323290-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR