5-Bromo-2-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-2-nitrophenyl)methanol

A409695-1G A409695-250MG A409695-5G
77102-008EA 45.62 USD
77102-008 77102-010 77102-012
5-Bromo-2-nitrobenzyl alcohol 98%
5-Bromo-2-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.031 g/mol
MDL Number: MFCD11848688
CAS Number: 1241894-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR