L-Tyrosine amide 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr-NH₂, H-Tyr-NH2

A409561-1G A409561-5G
77102-006EA 29 USD
77102-006 77686-130
L-Tyrosine amide 98%
L-Tyrosine amide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD00069935
CAS Number: 4985-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR