5-(tert-Butyl)-1H-pyrrole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A409506-100MG A409506-1G A409506-250MG
77102-000EA 24.41 USD
77102-000 77102-002 77102-004
5-(tert-Butyl)-1H-pyrrole-2-carbaldehyde 95%
5-(tert-Butyl)-1H-pyrrole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.20 g/mol
MDL Number: MFCD14705044
CAS Number: 59001-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR