Methyl 4-(2-fluoro-3-(3-(6-methylpyridin-3-yl)ureido)benzyl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Omecamtiv mecarbil

A408772-10MG A408772-1MG A408772-5MG
77101-990EA 41.38 USD
77101-990 77101-992 77101-994
Methyl 4-(2-fluoro-3-(3-(6-methylpyridin-3-yl)ureido)benzyl)piperazine-1-carboxylate 98%
Methyl 4-(2-fluoro-3-(3-(6-methylpyridin-3-yl)ureido)benzyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₄FN₅O₃
MW: 401.43 g/mol
MDL Number: MFCD18633260
CAS Number: 873697-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR