CZC-54252 hydrochloride 98%

Supplier: Ambeed
Warning

A408166-10MG A408166-1MG A408166-25MG A408166-50MG A408166-5MG
77101-980EA 50 USD
77101-980 77101-982 77101-984 77101-986 77101-988
CZC-54252 hydrochloride 98%
CZC-54252 hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₅ClN₆O₄S.HCl
MDL Number: MFCD26401527
CAS Number: 1191911-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR