1-(2-Amino-3,4-dimethoxyphenyl)ethanone 98+%
Supplier: Ambeed
|
Warning
|
A407670-100MG
A407670-1G
A407670-250MG
77101-974EA
68.96
USD
77101-974
77101-976
77101-978
1-(2-Amino-3,4-dimethoxyphenyl)ethanone 98+%
1-(2-Amino-3,4-dimethoxyphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₃ MW: 195.2151 g/mol |
MDL Number:
MFCD11040634 CAS Number: 49701-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...