1-(2-Hydroxyphenyl)butan-1-one 95%

Supplier: Ambeed
Warning

A405519-100G A405519-5G
77101-956EA 114.58 USD
77101-956 77101-958
1-(2-Hydroxyphenyl)butan-1-one 95%
1-(2-Hydroxyphenyl)butan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
MDL Number: MFCD00020041
CAS Number: 2887-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR