Ethyl 2-cyano-5-fluorobenzoate 95+%

Supplier: Ambeed
Warning

A405162-1G A405162-5G
77101-952EA 33 USD
77101-952 77101-954
Ethyl 2-cyano-5-fluorobenzoate 95+%
Ethyl 2-cyano-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MDL Number: MFCD11041402
CAS Number: 1260751-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR