(R)-4-Benzyl-5,5-dimethyloxazolidin-2-one 98%

Supplier: Ambeed
Warning

A404627-100MG A404627-1G A404627-250MG
77101-946EA 20 USD
77101-946 77101-948 77101-950
(R)-4-Benzyl-5,5-dimethyloxazolidin-2-one 98%
(R)-4-Benzyl-5,5-dimethyloxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MDL Number: MFCD00274260
CAS Number: 204851-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR