Methyl pyrazolo[1,5-a]pyridine-5-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A404086-1G
A404086-250MG
77101-942EA
68.96
USD
77101-942
77686-112
Methyl pyrazolo[1,5-a]pyridine-5-carboxylate 98%
Methyl pyrazolo[1,5-a]pyridine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₈N₂O₂ MW: 176.1720 g/mol |
MDL Number:
MFCD21364554 CAS Number: 1101120-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...