Chloromethyl pivalate 99%

Supplier: Ambeed
Danger

Synonyms: Chloromethyl trimethylacetate, Trimethylacetic acid chloromethyl ester, 2,2-Dimethylpropionic acid chloromethyl ester, Pivalic acid chloromethyl ester

A403326-25G A403326-5G
77101-938EA 6.37 USD
77101-938 77101-940
Chloromethyl pivalate 99%
Chloromethyl pivalate

Precaución: for Research Use Only

Formula: C₆H₁₁ClO₂
MW: 150.6 g/mol
MDL Number: MFCD00000884
CAS Number: 18997-19-8
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR