9,10-Bis(4-methoxyphenyl)anthracene 98%

Supplier: Ambeed
Warning

A402838-1G A402838-25G A402838-5G
77101-932EA 11.67 USD
77101-932 77101-934 77101-936
9,10-Bis(4-methoxyphenyl)anthracene 98%
9,10-Bis(4-methoxyphenyl)anthracene

Precaución: for Research Use Only

Formula: C₂₈H₂₂O₂
MW: 390.48 g/mol
MDL Number: MFCD00009936
CAS Number: 24672-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR