Ethyl 4-oxo-1,4-dihydroquinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

A402663-1G A402663-250MG A402663-25G A402663-5G
77101-924EA 20 USD
77101-924 77101-926 77101-928 77101-930
Ethyl 4-oxo-1,4-dihydroquinoline-3-carboxylate 95%
Ethyl 4-oxo-1,4-dihydroquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₃
MW: 217.221 g/mol
MDL Number: MFCD01314279
CAS Number: 52980-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR