Ethyl-trans-3-(3-pyridyl)acrylate 98%

Supplier: Ambeed
Warning

A402475-1G A402475-25G A402475-5G
77101-918EA 32.89 USD
77101-918 77101-920 77101-922
Ethyl-trans-3-(3-pyridyl)acrylate 98%
Ethyl-trans-3-(3-pyridyl)acrylate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.20 g/mol
MDL Number: MFCD02177443
CAS Number: 59607-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR