1-(Naphthalen-1-yl)ethanone oxime 95+%

Supplier: Ambeed
Danger

A402011-1G A402011-250MG A402011-5G
77101-912EA 76 USD
77101-912 77101-914 77101-916
1-(Naphthalen-1-yl)ethanone oxime 95+%
1-(Naphthalen-1-yl)ethanone oxime

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MDL Number: MFCD00550305
CAS Number: 1956-40-7
UN: 2926
ADR: 4.1,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR