5-((tert-Butoxycarbonyl)amino)-2-fluorobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A400777-100MG
A400777-5G
77101-908EA
28.65
USD
77101-908
77572-240
5-((tert-Butoxycarbonyl)amino)-2-fluorobenzoic acid 95%
5-((tert-Butoxycarbonyl)amino)-2-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₄FNO₄ MW: 255.24 g/mol |
MDL Number:
MFCD12525335 CAS Number: 141940-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...