Ethyl [(tert-butoxycarbonyl)amino](oxo)acetate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Ethyl 2-((tert-butoxycarbonyl)amino)-2-oxoacetate
A400393-250MG
A400393-1G
A400393-5G
77101-906EA
18
USD
77101-906
77686-106
77900-832
Ethyl [(tert-butoxycarbonyl)amino](oxo)acetate 95%
Ethyl [(tert-butoxycarbonyl)amino](oxo)acetate
Precaución: for Research Use Only
Formula:
C₉H₁₅NO₅ |
MDL Number:
MFCD01631201 CAS Number: 216959-34-1 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...