2-(4-(Bromomethyl)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: Ambeed
Danger

A400346-1G A400346-250MG A400346-5G
77101-900EA 157.02 USD
77101-900 77101-902 77101-904
2-(4-(Bromomethyl)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(4-(Bromomethyl)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₇H₂₀BBrO₂
MW: 347.05 g/mol
MDL Number: MFCD20441898
CAS Number: 1422655-33-1
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR