XL888 95+%

Supplier: Ambeed
Warning

Synonyms: 5-(((R)-sec-Butyl)amino)-N1-((3-endo)-8-(5-(cyclopropanecarbonyl)pyridin-2-yl)-8-azabicyclo[3.2.1]octan-3-yl)-2-methylterephthalamide

A399992-10MG A399992-1MG A399992-25MG A399992-50MG A399992-5MG
77101-890EA 83.82 USD
77101-890 77101-892 77101-894 77101-896 77101-898
XL888 95+%
XL888

Precaución: for Research Use Only

Formula: C₂₉H₃₇N₅O₃
MW: 503.64 g/mol
MDL Number: MFCD22124888
CAS Number: 1149705-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR