tert-Butyl 2,5-diazabicyclo[4.1.0]heptane-2-carboxylate 98%

Supplier: Ambeed
Warning

A398994-100MG A398994-250MG A398994-1G
77101-880EA 36 USD
77101-880 77101-882 77568-808
tert-Butyl 2,5-diazabicyclo[4.1.0]heptane-2-carboxylate 98%
tert-Butyl 2,5-diazabicyclo[4.1.0]heptane-2-carboxylate

Precaución: For research use only.

Formula: C₁₀H₁₈N₂O₂
MDL Number: MFCD17480504
CAS Number: 1228675-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR