1-(Dimethylamino)-2-methylpropan-2-ol 97%

Supplier: Ambeed
Warning

A398548-1G A398548-250MG A398548-25G
77101-874EA 66.84 USD
77101-874 77101-876 77572-230
1-(Dimethylamino)-2-methylpropan-2-ol 97%
1-(Dimethylamino)-2-methylpropan-2-ol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MW: 117.189 g/mol
MDL Number: MFCD11934288
CAS Number: 14123-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR