3'-Methoxybiphenyl-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3'-Methoxy-[1,1'-biphenyl]-3-carboxylic acid

A398164-100MG A398164-1G A398164-250MG A398164-5G
77101-862EA 14.86 USD
77101-862 77101-864 77101-866 77101-868
3'-Methoxybiphenyl-3-carboxylic acid 98%
3'-Methoxybiphenyl-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD03424598
CAS Number: 168618-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR