2-(Trimethylsilyl)ethanol 98%

Supplier: Ambeed
Danger

A398015-5G A398015-25G
77101-856EA 14.99 USD
77101-856 77551-190
2-(Trimethylsilyl)ethanol 98%
2-(Trimethylsilyl)ethanol

Precaución: for Research Use Only

Formula: HOH₂CCH₂Si(CH₃)₃
MDL Number: MFCD00002825
CAS Number: 2916-68-9
UN: 1987
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR