(S)-2-((tert-Butoxycarbonyl)amino)-3-(2-fluoro-4-hydroxyphenyl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Tyr(2-F)-OH

A394681-1G A394681-250MG
77101-830EA 457 USD
77101-830 77101-832
(S)-2-((tert-Butoxycarbonyl)amino)-3-(2-fluoro-4-hydroxyphenyl)propanoic acid 95%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(2-fluoro-4-hydroxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₅
MW: 299.29 g/mol
MDL Number: MFCD14585164
CAS Number: 125218-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR