3-(4-Fluorophenoxy)phenylboronic acid 95%

Supplier: Ambeed
Warning

A394627-100MG A394627-1G A394627-250MG A394627-5G
77101-822EA 39 USD
77101-822 77101-824 77101-826 77101-828
3-(4-Fluorophenoxy)phenylboronic acid 95%
3-(4-Fluorophenoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BFO₃
MDL Number: MFCD14584726
CAS Number: 283173-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR