2,3-Dibromo-5-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dibromo-5-methylpyridine

A394100-25G A394100-5G
77101-818EA 17 USD
77101-818 77101-820
2,3-Dibromo-5-picoline 98%
2,3-Dibromo-5-picoline

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.92
MDL Number: MFCD04112574
CAS Number: 29232-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR