2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A392598-1G A392598-250MG A392598-5G
77101-808EA 210 USD
77101-808 77101-810 77101-812
2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 98%
2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₅BClNO₂
MDL Number: MFCD18729917
CAS Number: 1449475-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR