1-Chloro-2,6-difluorobenzene 98%

Supplier: Ambeed
Danger

A392358-25G A392358-5G
77101-806EA 31 USD
77101-806 77686-085
1-Chloro-2,6-difluorobenzene 98%
1-Chloro-2,6-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClF₂
MDL Number: MFCD00142604
CAS Number: 38361-37-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR