4-(Cyclopropylmethoxy)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Cyclopropylmethoxy)phenyl)boronic acid, 4-(Cyclopropylmethoxy)phenylboronic acid

A392332-1G A392332-250MG A392332-5G
77101-800EA 107 USD
77101-800 77101-802 77101-804
4-(Cyclopropylmethoxy)benzeneboronic acid 98%
4-(Cyclopropylmethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₃
MDL Number: MFCD04039017
CAS Number: 411229-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR