L(+)-Cysteine 97%

Supplier: Ambeed
Warning

Synonyms: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

A390912-1000G A390912-100G A390912-10G A390912-25G A390912-500G
77101-778EA 71 USD
77101-778 77101-780 77101-782 77101-784 77101-786
L(+)-Cysteine 97%
L-Cysteine

Precaución: for Research Use Only

Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
Boiling Pt: 254
Melting Pt: 115
MDL Number: MFCD00064306
CAS Number: 52-90-4
Merck Index: 13,02810

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR