4,4'-Difluorobenzhydryl chloride 98%

Supplier: Ambeed
Danger

Synonyms: Chloro bis-(4-fluorophenyl)methane, 4,4-Difluorodiphenylmethylchloride

A390539-1G A390539-5G A390539-25G
77101-764EA 7.43 USD
77101-764 77551-156 77594-190
4,4'-Difluorobenzhydryl chloride 98%
4,4'-Difluorobenzhydryl chloride

Precaución: for Research Use Only

Formula: C₁₃H₉ClF₂
MW: 238.66 g/mol
MDL Number: MFCD00044329
CAS Number: 27064-94-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR