UNC0638 98%

Supplier: Ambeed
Warning

A389386-10MG A389386-1MG A389386-5MG
77101-754EA 47 USD
77101-754 77101-756 77101-758
UNC0638 98%
UNC0638

Precaución: for Research Use Only

Formula: C₃₀H₄₇N₅O₂
MDL Number: MFCD22208587
CAS Number: 1255580-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR