4-(3-Bromophenyl)-2,6-diphenylpyrimidine 97%

Supplier: Ambeed
Warning

A389145-1G A389145-5G
77101-750EA 11.67 USD
77101-750 77101-752
4-(3-Bromophenyl)-2,6-diphenylpyrimidine 97%
4-(3-Bromophenyl)-2,6-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₂₂H₁₅BrN₂
MW: 387.28 g/mol
MDL Number: MFCD28962150
CAS Number: 864377-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR