Z-L-Aspartic acid α-tert-butyl ester dicyclohexylammonium salt 98%

Supplier: Ambeed
Warning

Synonyms: Z-Asp-OtBu·DCHA, Dicyclohexylamine (S)-2-(((benzyloxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoate, Z-Asp(OtBu)-OH·DCHA

A385316-1G A385316-25G A385316-5G
77101-742EA 21 USD
77101-742 77101-744 77101-746
Z-L-Aspartic acid α-tert-butyl ester dicyclohexylammonium salt 98%
Z-L-Aspartic acid α-tert-butyl ester dicyclohexylammonium salt

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₆·C₁₂H₂₃N
MW: 504.668 g/mol
MDL Number: MFCD00038885
CAS Number: 23632-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR