2-Mercaptopyridine-N-oxide sodium salt 96%

Supplier: Ambeed
Danger

Synonyms: 2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium

A384633-25G A384633-100G A384633-500G
77101-740EA 19.1 USD
77101-740 77613-142 77613-144
2-Mercaptopyridine-N-oxide sodium salt 96%
2-Mercaptopyridine N-oxide sodium salt

Precaución: for Research Use Only

Formula: C₅H₄NNaOS
MW: 149.15 g/mol
MDL Number: MFCD01941547
CAS Number: 3811-73-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR