(2-Pyridyl)acetic acid ethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-Pyridylacetate, Ethyl 2-pyridineacetate, Ethyl (2-pyridyl)acetate

A384401-100G A384401-10G A384401-1G A384401-25G A384401-5G
77101-730EA 398.9 USD
77101-730 77101-732 77101-734 77101-736 77101-738
(2-Pyridyl)acetic acid ethyl ester 95%
(2-Pyridyl)acetic acid ethyl ester

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00006359
CAS Number: 2739-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR