2-Chloro-6-(trifluoromethyl)nicotinamide 97%

Supplier: Ambeed
Warning

A383467-100MG A383467-250MG
77101-718EA 8.49 USD
77101-718 77101-720
2-Chloro-6-(trifluoromethyl)nicotinamide 97%
2-Chloro-6-(trifluoromethyl)nicotinamide

Precaución: for Research Use Only

Formula: C₇H₄ClF₃N₂O
MDL Number: MFCD01862659
CAS Number: 386704-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR